Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalin 98.0+%, TCI America™
SDP

Catalog No. P009525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P009525G 25 g
P0095500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P009525G Supplier TCI America Supplier No. P009525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81-90-3
Molecular Formula C20H16O4
Molecular Weight (g/mol) 320.344
MDL Number MFCD00036135
InChI Key FFFPYJTVNSSLBQ-UHFFFAOYSA-N
PubChem CID 66494
ChEBI CHEBI:34915
IUPAC Name 2-[bis(4-hydroxyphenyl)methyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C(=O)O

Specifications

Melting Point 235°C
Formula Weight 320.34
Percent Purity ≥98.0% (HPLC,T)
Color White
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Phenolphthalin [for redox indicator]
EINECSNumber (4)-0263
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.