Learn More
Chem Service Inc PHENOLS MIX #9 2000UG/ML
SDP
Catalog No. NC0721246
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Mixture #9-Phenols, 2000ug/ml in Methylene chloride, Size: 1mL Ampule
Catalog No. NC0721246 Supplier Chem Service Inc Supplier No. MPPHC9X51ML
This item has a minimum order quantity of 2 per supplier requirements
Mixture #9-Phenols, 2000ug/ml in Methylene chloride, Size: 1mL Ampule