Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Phenomenex bioZen SEC Column, 1.8 μm, 50 x 2.1 mm

Catalog No. NC3842361 Shop All Phenomenex Products
Encompass

The Phenomenex bioZen SEC-2 1.8 μm LC column (50 x 2.1 mm) is a UHPLC-compatible size-exclusion chromatography column optimized for low molecular weight separations (1 k-450 kDa). It employs high-strength 1.8 μm porous particles and a 150 Å pore size to improve resolution and sensitivity for peptides, protein fragments, and small biologics, while supporting a broad pH range and HPLC system pressures.

  • 1.8 μm particle size.
  • Phase: SEC-2.
  • Dimensions: 50 x 2.1 mm (length x internal diameter).
  • Pore size: 150 Å.
  • Recommended pH range: 1.5-8.5.
  • Low molecular weight separation range: 1 k-450 kDa.
  • Maximum backpressure: 480 bar.
  • Shipping solvent: 0.1 M phosphate buffer, pH 6.8, with 0.025% NaN3.

Catalog No. NC3842361 Supplier Phenomenex Inc Supplier No. PART 00B4769AN
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The Phenomenex bioZen SEC-2 1.8 μm LC column (50 x 2.1 mm) is a UHPLC-compatible size-exclusion chromatography column optimized for low molecular weight separations (1 k-450 kDa). It employs high-strength 1.8 μm porous particles and a 150 Å pore size to improve resolution and sensitivity for peptides, protein fragments, and small biologics, while supporting a broad pH range and HPLC system pressures.

  • 1.8 μm particle size.
  • Phase: SEC-2.
  • Dimensions: 50 x 2.1 mm (length x internal diameter).
  • Pore size: 150 Å.
  • Recommended pH range: 1.5-8.5.
  • Low molecular weight separation range: 1 k-450 kDa.
  • Maximum backpressure: 480 bar.
  • Shipping solvent: 0.1 M phosphate buffer, pH 6.8, with 0.025% NaN3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.