Learn More
Phenomenex Inc Phenomenex Clarity OTX Pro Extraction Kit, 100 mg, 96-Well
Catalog No. NC3919800 Shop All Phenomenex Products
Encompass
The Clarity OTX Pro Extraction Kit provides a 96-well solid phase extraction workflow using 100 mg sorbent per well for cleanup and isolation of nucleic acid-based therapeutics and small-molecule metabolites from biological matrices. The kit includes a prepacked 96-well plate and matching buffers to support high-throughput bioanalytical sample preparation.
  • 96-well plate format.
  • 100 mg sorbent per well.
  • Ion exchange sorbent chemistry.
  • Includes extraction buffers for conditioning and elution.
  • Suitable for RNA and DNA therapeutics and metabolite extraction.
  • Designed for high-throughput bioanalytical workflows.
Catalog No. NC3919800 Supplier Phenomenex Inc Supplier No. KS09540
The Clarity OTX Pro Extraction Kit provides a 96-well solid phase extraction workflow using 100 mg sorbent per well for cleanup and isolation of nucleic acid-based therapeutics and small-molecule metabolites from biological matrices. The kit includes a prepacked 96-well plate and matching buffers to support high-throughput bioanalytical sample preparation.
  • 96-well plate format.
  • 100 mg sorbent per well.
  • Ion exchange sorbent chemistry.
  • Includes extraction buffers for conditioning and elution.
  • Suitable for RNA and DNA therapeutics and metabolite extraction.
  • Designed for high-throughput bioanalytical workflows.