Learn More
Phenomenex Inc Phenomenex Clarity OTX SPE Plate, 25 mg/well, 96-Well
Catalog No. NC3919793 Shop All Phenomenex Products
Encompass
The Clarity OTX 25 mg 96-well SPE plate provides OTX sorbent chemistry with 25 mg sorbent per well in a 96-well format. It is designed for high-capacity extraction and cleanup of oligonucleotide therapeutics from biological matrices and is compatible with automated liquid handlers for streamlined sample preparation prior to LC-MS analysis.
  • 25 mg sorbent per well for high-capacity extraction.
  • OTX sorbent chemistry tailored for oligonucleotide retention and cleanup.
  • 96-well format compatible with automated liquid handling systems.
  • Optimized for extraction of phosphorothioate and conjugated oligonucleotides prior to LC-MS.
  • Single plate per box for direct workflow integration.
Catalog No. NC3919793 Supplier Phenomenex Inc Supplier No. 8ES103DGA
Add to Cart
Add to Cart
The Clarity OTX 25 mg 96-well SPE plate provides OTX sorbent chemistry with 25 mg sorbent per well in a 96-well format. It is designed for high-capacity extraction and cleanup of oligonucleotide therapeutics from biological matrices and is compatible with automated liquid handlers for streamlined sample preparation prior to LC-MS analysis.
  • 25 mg sorbent per well for high-capacity extraction.
  • OTX sorbent chemistry tailored for oligonucleotide retention and cleanup.
  • 96-well format compatible with automated liquid handling systems.
  • Optimized for extraction of phosphorothioate and conjugated oligonucleotides prior to LC-MS.
  • Single plate per box for direct workflow integration.