Learn More
Phenomenex Inc Phenomenex GC Inlet Liner, 4 mm ID, Single Taper, Splitless
Catalog No. NC3732075 Shop All Phenomenex Products
Encompass
This Phenomenex GC inlet liner is engineered for splitless injections in Agilent-style split/splitless inlet systems. The single-taper geometry and 4 mm inner diameter promote sample focusing and efficient transfer for trace-level analyses. Supplied in multi-packs for routine replacement and maintenance.
  • 4 mm inner diameter
  • 78.5 mm length
  • 6.45 mm outer diameter
  • Single taper geometry for focused injections
  • Designed for splitless injection applications
  • Packaged 25 per pack
Catalog No. NC3732075 Supplier Phenomenex Inc Supplier No. AG07640
This Phenomenex GC inlet liner is engineered for splitless injections in Agilent-style split/splitless inlet systems. The single-taper geometry and 4 mm inner diameter promote sample focusing and efficient transfer for trace-level analyses. Supplied in multi-packs for routine replacement and maintenance.
  • 4 mm inner diameter
  • 78.5 mm length
  • 6.45 mm outer diameter
  • Single taper geometry for focused injections
  • Designed for splitless injection applications
  • Packaged 25 per pack