Learn More
Phenomenex Inc Phenomenex Lysis Loading Buffer, OTX Pro, 1 L, DNA/RNA Extraction
Catalog No. NC3919794 Shop All Phenomenex Products
Encompass
Lysis-loading buffer formulated for efficient extraction of RNA- and DNA-based therapeutics and their metabolites from biological matrices. Supplied as a 1 L solution, the buffer provides cell lysis and protease inactivation and conditions samples for ion-exchange solid phase extraction workflows.
  • Supplied as a 1 L solution.
  • Formulated for extraction of RNA- and DNA-based therapeutics and their metabolites.
  • Provides cell lysis and removes protease activity in biological fluids.
  • Compatible with ion-exchange solid phase extraction workflows.
Catalog No. NC3919794 Supplier Phenomenex Inc Supplier No. AL08500
Lysis-loading buffer formulated for efficient extraction of RNA- and DNA-based therapeutics and their metabolites from biological matrices. Supplied as a 1 L solution, the buffer provides cell lysis and protease inactivation and conditions samples for ion-exchange solid phase extraction workflows.
  • Supplied as a 1 L solution.
  • Formulated for extraction of RNA- and DNA-based therapeutics and their metabolites.
  • Provides cell lysis and removes protease activity in biological fluids.
  • Compatible with ion-exchange solid phase extraction workflows.