Learn More
Phenomenex Inc Phenomenex Verex Vial, 9 mm Screw, Polypropylene, 300 μL
Catalog No. NC3812016 Shop All Phenomenex Products
Encompass_Preferred
The Verex 9 mm screw-top polypropylene vial provides a compact 300 μL nominal capacity for sample preparation and storage, designed for use with autosamplers and chromatographic workflows.
  • 300 μL nominal capacity.
  • Polypropylene construction for chemical resistance.
  • 9 mm screw-top opening.
  • Clear vial for visual sample inspection.
  • Supplied 100 per pack.
  • Compatible with standard autosamplers and vial racks.
Catalog No. NC3812016 Supplier Phenomenex Inc Supplier No. AR039P212
The Verex 9 mm screw-top polypropylene vial provides a compact 300 μL nominal capacity for sample preparation and storage, designed for use with autosamplers and chromatographic workflows.
  • 300 μL nominal capacity.
  • Polypropylene construction for chemical resistance.
  • 9 mm screw-top opening.
  • Clear vial for visual sample inspection.
  • Supplied 100 per pack.
  • Compatible with standard autosamplers and vial racks.