Learn More
Revvity Health Sciences Inc PhenoPlate 384-well, black, optically clear flat-bottom, fibronectin-coated, lids, case of 40
Catalog No. 502809582
Encompass
PhenoPlate 384-well, black, optically clear flat-bottom, fibronectin-coated, lids, case of 40
Catalog No. 50-280-9582 Supplier Revvity Health Sciences Inc Supplier No. 6057602
PhenoPlate 384-well, black, optically clear flat-bottom, fibronectin-coated, lids, case of 40