Learn More
Medchemexpress LLC Phenothiazin-5-ium, 3,7-bis(dimethylamino)-, chloride, hydrate | 122965-43-9 | 100 G
SDP
Catalog No. 5000409937
Encompass_Preferred
Methylene blue hydrate is an inhibitor of guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS). It acts as a vasopressor and is commonly used as a dye in various medical procedures. This compound influences the nitric oxide synthase/guanylate cyclase signaling pathway to decrease prepulse inhibition.
  • Acts as a REDOX cycling compound
  • Capable of crossing the blood-brain barrier
  • Identified as a Tau aggregation inhibitor
  • Reduces cerebral edema and attenuates microglial activation
  • Lessens neuroinflammation
Catalog No. 50-004-09937 Supplier Medchemexpress LLC Supplier No. HYD0958100G
Methylene blue hydrate is an inhibitor of guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS). It acts as a vasopressor and is commonly used as a dye in various medical procedures. This compound influences the nitric oxide synthase/guanylate cyclase signaling pathway to decrease prepulse inhibition.
  • Acts as a REDOX cycling compound
  • Capable of crossing the blood-brain barrier
  • Identified as a Tau aggregation inhibitor
  • Reduces cerebral edema and attenuates microglial activation
  • Lessens neuroinflammation