Learn More
Selleck Chemical LLC Phenothiazine S4251-50mg
SDP
Catalog No. 503108383
Encompass_Preferred
Phenothiazine (ENT 38) is a dopamine-2 (D2) receptor antagonist therefore decreases the effect of dopamine in the brain
Catalog No. 50-310-8383 Supplier Selleck Chemical LLC Supplier No. S425150MG
Phenothiazine (ENT 38) is a dopamine-2 (D2) receptor antagonist therefore decreases the effect of dopamine in the brain