Learn More
Medchemexpress LLC HY-Y0055 500mg Medchemexpress, Phenothiazine CAS:92-84-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0055500MG
Encompass_Preferred
Medchemexpress, HY-Y0055 500mg Phenothiazine CAS:92-84-2 Phenothiazine is an antibiotic which has insecticidal, fungicidal, antibacterial and anthelmintic activities. Phenothiazine also can be used for the research of neurological diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3516