Learn More
Apexbio Technology LLC Phenothiazine 92-84-2 50mg
SDP
Catalog No. 502240704
Encompass_Preferred
Phenothiazine (CAS 92-84-2) is a small-molecule antagonist targeting the dopamine-2 (D2) receptor It is designed to inhibit D2 receptor activity thereby reducing dopaminergic neurotransmission within neural pathways Phenothiazine exerts its biological activity primarily through direct binding to and inhibition of the D2 receptor In cellular and animal studies phenothiazine demonstrates receptor antagonism with an IC50 value generally reported in the micromolar range Based on these pharmacological properties phenothiazine holds research potential in neuroscience particularly for studies on dopamine-dependent pathways and related neurological disorders
This item is not returnable. View return policy
Catalog No. 50-224-0704 Supplier Apexbio Technology LLC Supplier No. B181550
Phenothiazine (CAS 92-84-2) is a small-molecule antagonist targeting the dopamine-2 (D2) receptor It is designed to inhibit D2 receptor activity thereby reducing dopaminergic neurotransmission within neural pathways Phenothiazine exerts its biological activity primarily through direct binding to and inhibition of the D2 receptor In cellular and animal studies phenothiazine demonstrates receptor antagonism with an IC50 value generally reported in the micromolar range Based on these pharmacological properties phenothiazine holds research potential in neuroscience particularly for studies on dopamine-dependent pathways and related neurological disorders