Learn More
Medchemexpress LLC Phenothiazine-d8 | 1219803-41-4 | 99.0% | 207.32 | C12HD8NS | 10mg
SDP
Catalog No. 5000268271
Encompass_Preferred
Phenothiazine-d8 is the deuterium labeled Phenothiazine Phenothiazine is an antibiotic which has insecticidal fungicidal antibacterial and anthelmintic activities Phenothiazine also can be used for the research of neurological diseases[1][2]
Catalog No. 50-002-68271 Supplier Medchemexpress LLC Supplier No. HYY0055S10MG
Phenothiazine-d8 is the deuterium labeled Phenothiazine Phenothiazine is an antibiotic which has insecticidal fungicidal antibacterial and anthelmintic activities Phenothiazine also can be used for the research of neurological diseases[1][2]