Learn More
Medchemexpress LLC Phenothrin | 26002-80-2 | 97.7% | 5 MG
SDP
Catalog No. 5000413635
Encompass_Preferred
Phenothrin is a Type I pyrethroid insecticide that induces dose-dependent DNA damage in human peripheral blood lymphocytes and hepatocytes. It exhibits genotoxic potential and is used for pest control in agriculture, households, public health, and for eliminating human head lice infestations.
  • Induces dose-dependent DNA damage
  • Exhibits genotoxic potential
  • Used for pest control in agriculture, households, and public health
  • Effective for eliminating human head lice infestations
  • Available in liquid form
Catalog No. 50-004-13635 Supplier Medchemexpress LLC Supplier No. HYB10725MG
Phenothrin is a Type I pyrethroid insecticide that induces dose-dependent DNA damage in human peripheral blood lymphocytes and hepatocytes. It exhibits genotoxic potential and is used for pest control in agriculture, households, public health, and for eliminating human head lice infestations.
  • Induces dose-dependent DNA damage
  • Exhibits genotoxic potential
  • Used for pest control in agriculture, households, and public health
  • Effective for eliminating human head lice infestations
  • Available in liquid form