Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenothrin | 26002-80-2 | MFCD00078716 | 96.0% | 350.45 | C23H26O3 | 10 MG
SDP

Catalog No. 5000297036
Encompass_Preferred

Phenothrin is a synthetic pyrethroid insecticide supplied as a research-grade standard for laboratory use. It is used as a parasite inhibitor in studies of insecticidal activity against fleas, ticks, and lice, and is provided as a small 10 mg liquid sample suitable for analytical and laboratory applications.

  • Synthetic pyrethroid insecticide used as a parasite inhibitor.
  • Provided as a 10 mg liquid research standard.
  • CAS number 26002-80-2; molecular formula C23H26O3; molecular weight 350.45.
  • Purity approximately 96.0%.
  • Intended for research and analytical applications only.
  • Storage recommendations: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-97036 Supplier Medchemexpress LLC Supplier No. HYB107210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenothrin is a synthetic pyrethroid insecticide supplied as a research-grade standard for laboratory use. It is used as a parasite inhibitor in studies of insecticidal activity against fleas, ticks, and lice, and is provided as a small 10 mg liquid sample suitable for analytical and laboratory applications.

  • Synthetic pyrethroid insecticide used as a parasite inhibitor.
  • Provided as a 10 mg liquid research standard.
  • CAS number 26002-80-2; molecular formula C23H26O3; molecular weight 350.45.
  • Purity approximately 96.0%.
  • Intended for research and analytical applications only.
  • Storage recommendations: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.