Learn More
Revvity Health Sciences Inc Phosphorus-32 Radionuclide, 1mCi (37MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water
Catalog No. 502812361
Encompass
Phosphorus-32 Radionuclide, 1mCi (37MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water
Catalog No. 50-281-2361 Supplier Revvity Health Sciences Inc Supplier No. NEX053001MC
Phosphorus-32 Radionuclide, 1mCi (37MBq), Specific Activity: 8500-9120Ci (314-337TBq)/mmol, Orthophosphoric Acid in 1mL Water