Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Phenoxathiin | ≥97% | CAS 262-20-4 | C12H8OS | 25g

Catalog No. 5001040722
Encompass_Preferred

Phenoxathiin | ≥97% | CAS 262-20-4 | C12H8OS | 25g

Catalog No. 50-010-40722 Supplier Aobchem Supplier No. A2866525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenoxathiin | ≥97% | CAS 262-20-4 | C12H8OS | 25g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.