Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxazine, 98%

Catalog No. AAL0196903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0196903 1 g
AAL0196906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0196903 Supplier Thermo Scientific Chemicals Supplier No. L0196903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 135-67-1 | C12H9NO | 183.21 g/mol

Phenoxazine is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenoxazine is used as primary and secondary intermediates.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-67-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00005014
InChI Key TZMSYXZUNZXBOL-UHFFFAOYSA-N
Synonym phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci
PubChem CID 67278
IUPAC Name 10H-phenoxazine
SMILES N1C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 154°C to 158°C
Quantity 1 g
Beilstein 143234
Merck Index 14,7253
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material Phenoxazine
EINECSNumber 205-210-8
RTECSNumber SP7450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.