Learn More
Medchemexpress LLC Phenoxyacetone 10mM 1mL | 621-87-4 | 150.17 g/mol | C9H10O2 | 1 ML
SDP
Catalog No. 5000308027
Encompass_Preferred
Phenoxyacetone is a small-molecule acetylcholinesterase (AChE) inhibitor supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research and enzymatic assays.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Suitable for in vitro acetylcholinesterase assays and biochemical screening.
  • High purity reported on accompanying certificate of analysis (COA).
  • Safety data sheet (SDS) and COA are available for documentation and compliance.
  • Also offered as solid quantities for preparation of custom stock solutions.
Catalog No. 50-003-08027 Supplier Medchemexpress LLC Supplier No. HY11679210MM 1ML
Phenoxyacetone is a small-molecule acetylcholinesterase (AChE) inhibitor supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research and enzymatic assays.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Suitable for in vitro acetylcholinesterase assays and biochemical screening.
  • High purity reported on accompanying certificate of analysis (COA).
  • Safety data sheet (SDS) and COA are available for documentation and compliance.
  • Also offered as solid quantities for preparation of custom stock solutions.