Learn More
Medchemexpress LLC Phenoxyacetone 1g | 621-87-4 | 150.18 g/mol | C9H10O2 | 1 G
SDP
Catalog No. 5000227530
Encompass_Preferred
Phenoxyacetone (1-phenoxy-2-propanone) is a small-molecule research chemical reported as a competitive acetylcholinesterase (AChE) inhibitor. It is supplied as a light yellow to yellow liquid and is accompanied by analytical documentation to support experimental use and safe handling.
  • Competitive acetylcholinesterase (AChE) inhibitor for biochemical research.
  • Molecular formula C9H10O2 and molecular weight 150.18 g/mol.
  • High purity (99.38%) confirmed by certificate of analysis.
  • Available in laboratory-scale sizes, including 1 g vials for small-scale experiments.
  • Provided with a safety data sheet for handling, storage, and hazard information.
Catalog No. 50-002-27530 Supplier Medchemexpress LLC Supplier No. HY1167921G
Phenoxyacetone (1-phenoxy-2-propanone) is a small-molecule research chemical reported as a competitive acetylcholinesterase (AChE) inhibitor. It is supplied as a light yellow to yellow liquid and is accompanied by analytical documentation to support experimental use and safe handling.
  • Competitive acetylcholinesterase (AChE) inhibitor for biochemical research.
  • Molecular formula C9H10O2 and molecular weight 150.18 g/mol.
  • High purity (99.38%) confirmed by certificate of analysis.
  • Available in laboratory-scale sizes, including 1 g vials for small-scale experiments.
  • Provided with a safety data sheet for handling, storage, and hazard information.