Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenoxyacetone 500mg | 621-87-4 | 150.18 g/mol | C9H10O2 | 500 MG
SDP

Catalog No. 5000230030
Encompass_Preferred

Phenoxyacetone (1-phenoxy-2-propanone, CAS 621-87-4) is a small-molecule research reagent that functions as a competitive acetylcholinesterase (AChE) inhibitor. It is supplied as a light yellow liquid with high purity for laboratory use; handle and store according to recommended conditions.

  • High purity (99.38%).
  • Molecular formula C9H10O2, molecular weight 150.18 g/mol.
  • Acts as a competitive acetylcholinesterase (AChE) inhibitor.
  • Soluble in DMSO (up to 200 mg/mL; sonication may be required).
  • Storage: pure form stable 3 years at -20°C and 2 years at 4°C; in solvent stable 6 months at -80°C.
  • Supplied in a 500 MG package; other sizes available by request.

Catalog No. 50-002-30030 Supplier Medchemexpress LLC Supplier No. HY116792500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenoxyacetone (1-phenoxy-2-propanone, CAS 621-87-4) is a small-molecule research reagent that functions as a competitive acetylcholinesterase (AChE) inhibitor. It is supplied as a light yellow liquid with high purity for laboratory use; handle and store according to recommended conditions.

  • High purity (99.38%).
  • Molecular formula C9H10O2, molecular weight 150.18 g/mol.
  • Acts as a competitive acetylcholinesterase (AChE) inhibitor.
  • Soluble in DMSO (up to 200 mg/mL; sonication may be required).
  • Storage: pure form stable 3 years at -20°C and 2 years at 4°C; in solvent stable 6 months at -80°C.
  • Supplied in a 500 MG package; other sizes available by request.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.