Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenoxyacetyl chloride

Catalog No. 15862350G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-862-350G 50 g
15-862-3250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-862-350G Supplier Sigma Aldrich Supplier No. 15862350G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phenoxyacetyl chloride was used in the synthesis of:
  • Series of macrocyclic bis-α-lactams
  • 5-phenyl-6-epiphenoxymethylpenicillin benzyl ester
  • N-protected guanosine derivatives, useful in RNA synthesis
  • Phenyloxyketene, for cycloaddition to imines leading to α-lactams

Specifications

CAS 701-99-5
Density 1.235 g/mL (at 25°C (literature))
Boiling Point 225°C to 226°C (lit.)
Molecular Formula C8H7ClO2
Refractive Index n20/D 1.534 (literature)
Linear Formula C6H5OCH2COCl
MDL Number MFCD00000726
Quantity 50 g
Molecular Weight (g/mol) 170.59
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.