Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxyacetyl chloride, 98%

Catalog No. AC169942500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169942500 250 g Glass bottle
AC169940100 10 g Glass bottle
AC169940500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC169942500 Supplier Thermo Scientific Chemicals Supplier No. 169942500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 701-99-5 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 701-99-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000726
InChI Key PKUPAJQAJXVUEK-UHFFFAOYSA-N
Synonym phenoxyacetyl chloride, acetyl chloride, phenoxy, phenyloxyacetyl chloride, phenoxyacetylchloride, phenoxyacetic acid chloride, acetyl chloride, 2-phenoxy, pac-cl, phenoxylacetylchloride, 2-phenoxyacetylchloride, phenoxy acetyl chloride
PubChem CID 69703
IUPAC Name 2-phenoxyacetyl chloride
SMILES ClC(=O)COC1=CC=CC=C1

Specifications

Color Yellow to Brown
Density 1.2300g/mL
Boiling Point 225°C to 226°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5324 to 1.5344
Linear Formula C6H5OCH2COCl
Quantity 250 g
Beilstein 06, 162
Specific Gravity 1.23
Solubility Information Solubility in water: reacts
Formula Weight 170.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenoxyacetyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 211-862-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.