Learn More
Apexbio Technology LLC Phenoxybenzamine HCl, 1g Cas# 63-92-3
SDP
Supplier:  Apexbio Technology LLC B13431000
Encompass_Preferred
Phenoxybenzamine HCl is a non-specific, irreversible alpha antagonist with long duration of action. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8533