Learn More
Apexbio Technology LLC Phenoxybenzamine HCl 63-92-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13435.1
Encompass_Preferred
Phenoxybenzamine HCl (CAS number 63-92-3) is an irreversible nonselective antagonist of alpha-adrenergic receptors It covalently binds to these receptors producing sustained blockage and therefore preventing receptor-mediated vasoconstriction Due to this prolonged alpha-receptor antagonism phenoxybenzamine hydrochloride is frequently utilized in biomedical research to study sympathetic nervous system functions receptor pharmacology and cardiovascular-related pathophysiology
Catalog No. 50-000-62622