Learn More
Selleck Chemical LLC Phenoxybenzamine HCl S2499-1g
SDP
Catalog No. 503112082
Encompass_Preferred
Phenoxybenzamine HCl (NSC 37448 NCI-c01661) is a non-specific irreversible alpha antagonist with an IC50 of 550 nM
Catalog No. 50-311-2082 Supplier Selleck Chemical LLC Supplier No. S24991G
Phenoxybenzamine HCl (NSC 37448 NCI-c01661) is a non-specific irreversible alpha antagonist with an IC50 of 550 nM