Learn More
Abcam Pheno x ybenzamine hydrochloride, calmodulin and alpha-adrenoceptor antagonist, 500MG
Catalog No. 502705560
Encompass_Preferred
MW 340.3 Da, Purity >95%. Irreversible calmodulin antagonist (IC₅₀ ~ 80 μM) and potent, nonselective, irreversible α-adrenoceptor antagonist (IC₅₀ = 550 nM). Antihypertensive and anti-inflammatory effects in vitro and in vivo. Cell-permeable. Blood-brain barrier impermeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5560 Supplier Abcam Supplier No. AB144619500MG
MW 340.3 Da, Purity >95%. Irreversible calmodulin antagonist (IC₅₀ ~ 80 μM) and potent, nonselective, irreversible α-adrenoceptor antagonist (IC₅₀ = 550 nM). Antihypertensive and anti-inflammatory effects in vitro and in vivo. Cell-permeable. Blood-brain barrier impermeable.
The product is subject to the following: Abcam Restricted Use Statement