Learn More
Medchemexpress LLC Phenoxybenzamine hydrochloride | 63-92-3 | 99.5% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0431A1G
Encompass_Preferred
Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist. It is primarily used in research for hypertension, particularly that caused by pheochromocytoma, and has also demonstrated antitumor activity.
  • Inhibits proliferation of U251 and U87MG cells.
  • Inhibits migration and invasion of U251 and U87MG cells.
  • Activates LINGO-1 and inhibits the TrkB-Akt pathway.
  • Prevents hippocampal cell death after oxygen glucose deprivation.
  • Demonstrates anti-tumorigenic effect in nude mice with U87MG tumors.
  • Exhibits neuroprotective properties in a rat model of severe traumatic brain injury.
Catalog No. 50-004-07854