Learn More
Medchemexpress LLC Phenoxybenzamine hydrochloride | 63-92-3 | 99.5% | 50 MG
SDP
Catalog No. 5000407043
Encompass_Preferred
Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist. It is commonly used for the research of hypertension, specifically that caused by pheochromocytoma. It also demonstrates antitumor activity.
  • Nonselective, irreversible, orally active α-adrenoceptor antagonist
  • Used for research on hypertension caused by pheochromocytoma
  • Demonstrates antitumor activity
Catalog No. 50-004-07043 Supplier Medchemexpress LLC Supplier No. HYB0431AR50MG
Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist. It is commonly used for the research of hypertension, specifically that caused by pheochromocytoma. It also demonstrates antitumor activity.
  • Nonselective, irreversible, orally active α-adrenoceptor antagonist
  • Used for research on hypertension caused by pheochromocytoma
  • Demonstrates antitumor activity