Learn More
Medchemexpress LLC Phenoxybenzamine hydrochloride | 63-92-3 | 99.5% | 500 MG
SDP
Catalog No. 5000405280
Encompass_Preferred
Phenoxybenzamine hydrochloride is a nonselective, irreversible, and orally active alpha-adrenoceptor antagonist. It is utilized in research concerning hypertension, particularly when caused by pheochromocytoma, and has demonstrated antitumor activity. This compound also exhibits significant biological activity in various in vitro and in vivo studies.
  • Nonselective and irreversible alpha-adrenoceptor antagonist
  • Orally active compound
  • Used in hypertension research for pheochromocytoma
  • Exhibits antitumor activity
  • Inhibits cell proliferation, migration, and invasion
  • Activates LINGO-1 and inhibits the TrkB-Akt pathway
  • Prevents hippocampal cell death after oxygen glucose deprivation
  • Shows anti-tumorigenic effects in tumor models
  • Provides neuroprotective benefits in traumatic brain injury models
Catalog No. 50-004-05280 Supplier Medchemexpress LLC Supplier No. HYB0431A500MG
Phenoxybenzamine hydrochloride is a nonselective, irreversible, and orally active alpha-adrenoceptor antagonist. It is utilized in research concerning hypertension, particularly when caused by pheochromocytoma, and has demonstrated antitumor activity. This compound also exhibits significant biological activity in various in vitro and in vivo studies.
  • Nonselective and irreversible alpha-adrenoceptor antagonist
  • Orally active compound
  • Used in hypertension research for pheochromocytoma
  • Exhibits antitumor activity
  • Inhibits cell proliferation, migration, and invasion
  • Activates LINGO-1 and inhibits the TrkB-Akt pathway
  • Prevents hippocampal cell death after oxygen glucose deprivation
  • Shows anti-tumorigenic effects in tumor models
  • Provides neuroprotective benefits in traumatic brain injury models