Learn More
Medchemexpress LLC Phenoxybenzamine hydrochloride | 63-92-3 | MFCD00055152 | 99.5% | 100 MG
SDP
Catalog No. 5000408943
Encompass_Preferred
Phenoxybenzamine hydrochloride (Standard) serves as the analytical standard for Phenoxybenzamine hydrochloride, intended for research and analytical applications. It is an irreversible, nonselective, and orally active α-adrenoceptor antagonist, frequently employed in hypertension research, particularly for cases caused by pheochromocytoma. This compound also exhibits antitumor activity.
  • Analytical standard for phenoxybenzamine hydrochloride
  • Intended for research and analytical applications
  • Nonselective α-adrenoceptor antagonist
  • Irreversible α-adrenoceptor antagonist
  • Orally active α-adrenoceptor antagonist
  • Used in research for hypertension caused by pheochromocytoma
  • Shows antitumor activity
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-08943 Supplier Medchemexpress LLC Supplier No. HYB0431AR100MG
Phenoxybenzamine hydrochloride (Standard) serves as the analytical standard for Phenoxybenzamine hydrochloride, intended for research and analytical applications. It is an irreversible, nonselective, and orally active α-adrenoceptor antagonist, frequently employed in hypertension research, particularly for cases caused by pheochromocytoma. This compound also exhibits antitumor activity.
  • Analytical standard for phenoxybenzamine hydrochloride
  • Intended for research and analytical applications
  • Nonselective α-adrenoceptor antagonist
  • Irreversible α-adrenoceptor antagonist
  • Orally active α-adrenoceptor antagonist
  • Used in research for hypertension caused by pheochromocytoma
  • Shows antitumor activity
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS