Learn More
Medchemexpress LLC Phenoxyethanol | 122-99-6 | 10MM/1ML
SDP
Catalog No. 5000411444
Encompass_Preferred
Phenoxyethanol is a broad-spectrum antimicrobial agent utilized in various applications, including cosmetics, vaccines, and textiles. It functions by acting as an uncoupling agent in oxidative phosphorylation and competitively inhibiting malate dehydrogenase.
  • Exhibits antimicrobial activity against gram-positive bacteria and fungi (C. albicans).
  • Enhances microbial uptake of low molecular weight substances.
  • Can be used for vaccine preparation.
  • Serves as a preservative in diverse products.
Catalog No. 50-004-11444 Supplier Medchemexpress LLC Supplier No. HYB172910MM/1ML
Phenoxyethanol is a broad-spectrum antimicrobial agent utilized in various applications, including cosmetics, vaccines, and textiles. It functions by acting as an uncoupling agent in oxidative phosphorylation and competitively inhibiting malate dehydrogenase.
  • Exhibits antimicrobial activity against gram-positive bacteria and fungi (C. albicans).
  • Enhances microbial uptake of low molecular weight substances.
  • Can be used for vaccine preparation.
  • Serves as a preservative in diverse products.