Learn More
Medchemexpress LLC Phenoxyethanol | 122-99-6 | 99.5% | 500 MG
SDP
Catalog No. 5000413182
Encompass_Preferred
Phenoxyethanol is a broad-spectrum antimicrobial agent. It acts as an uncouple agent in oxidative phosphorylation from respiration and competitively inhibits malate dehydrogenase. It is used as a preservative in cosmetics, vaccines, and textiles.
  • Acts as an uncouple agent in oxidative phosphorylation from respiration.
  • Competitively inhibits malate dehydrogenase.
  • Used as a preservative in cosmetics, vaccines, and textiles.
  • Exhibits antimicrobial activity against gram-positive bacteria and fungi C. albicans.
  • Enhances microbial uptake of low molecular weight substances and can be used for vaccine preparation.
Catalog No. 50-004-13182 Supplier Medchemexpress LLC Supplier No. HYB1729500MG
Phenoxyethanol is a broad-spectrum antimicrobial agent. It acts as an uncouple agent in oxidative phosphorylation from respiration and competitively inhibits malate dehydrogenase. It is used as a preservative in cosmetics, vaccines, and textiles.
  • Acts as an uncouple agent in oxidative phosphorylation from respiration.
  • Competitively inhibits malate dehydrogenase.
  • Used as a preservative in cosmetics, vaccines, and textiles.
  • Exhibits antimicrobial activity against gram-positive bacteria and fungi C. albicans.
  • Enhances microbial uptake of low molecular weight substances and can be used for vaccine preparation.