Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxyethanol, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18605108 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-108 120 mL Amber Glass Bottle
18-605-109 500 mL Amber Glass Bottle
18-605-110 1 L Amber Glass Bottle
18-605-111 4 L Amber Glass Bottle
18-605-112 4 x 4 L Amber Glass Bottle
18-605-113 20 L Amber Glass Bottle
18-605-114 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-605-108 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1077120ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1077, 122-99-6, C8H10O2

Spectrum™ Chemical Phenoxyethanol, Reagent , is a glycol ether and an organic chemical compound. This colorless oily liquid is often used in dermalogical products and often replaces sodium azide as a biological buffer. It can also be found as a preservative in cosmetics, vaccines and various pharmaceuticals. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
IUPAC Name 2-phenoxyethan-1-ol
SMILES OCCOC1=CC=CC=C1

Specifications

CAS Max % 1
Boiling Point 244 to 250°C
Packaging Amber Glass Bottle
Quantity 120 mL
Percent Purity 99%
Grade Reagent
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.