Learn More
Medchemexpress LLC Phenoxyethanol (Standard) | 122-99-6 | 100.0% | 50 MG
Supplier: Medchemexpress LLC HYB1729R50MG
Phenoxyethanol (Standard) serves as the analytical standard for Phenoxyethanol, designed for research and analytical applications. It functions as a broad-spectrum antimicrobial agent and acts as an uncoupling agent in oxidative phosphorylation. Additionally, it competitively inhibits malate dehydrogenase and is widely utilized as a preservative in various products including cosmetics, vaccines, and textiles.
- Intended for research and analytical applications.
- Exhibits broad-spectrum antimicrobial activity.
- Uncouples oxidative phosphorylation from respiration.
- Competitively inhibits malate dehydrogenase.
- Used as a preservative in cosmetics, vaccines, and textiles.
- Analytical standard for precise research.
- Commonly used in qualitative, quantitative, and methodological research experiments.