Learn More
Medchemexpress LLC Phenoxyethanol (Standard) | 122-99-6 | 99.98% | 250 MG
SDP
Catalog No. 5000414432
Encompass_Preferred
Phenoxyethanol (Standard) is the analytical standard of Phenoxyethanol. It is intended for research and analytical applications, and possesses a broad spectrum of antimicrobial activity. Phenoxyethanol acts as an uncoupling agent in oxidative phosphorylation from respiration and competitively inhibits malate dehydrogenase. It is utilized as a preservative in cosmetics, vaccines, and textiles, among other applications. The compound is also used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for phenoxyethanol.
  • Broad spectrum antimicrobial agent.
  • Uncouples oxidative phosphorylation from respiration.
  • Competitively inhibits malate dehydrogenase.
  • Used as a preservative in cosmetics, vaccines, and textiles.
  • Suitable for qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
Catalog No. 50-004-14432 Supplier Medchemexpress LLC Supplier No. 250MGHYB1729R
Phenoxyethanol (Standard) is the analytical standard of Phenoxyethanol. It is intended for research and analytical applications, and possesses a broad spectrum of antimicrobial activity. Phenoxyethanol acts as an uncoupling agent in oxidative phosphorylation from respiration and competitively inhibits malate dehydrogenase. It is utilized as a preservative in cosmetics, vaccines, and textiles, among other applications. The compound is also used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for phenoxyethanol.
  • Broad spectrum antimicrobial agent.
  • Uncouples oxidative phosphorylation from respiration.
  • Competitively inhibits malate dehydrogenase.
  • Used as a preservative in cosmetics, vaccines, and textiles.
  • Suitable for qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).