Learn More
Cayman Chemical Phenprocoumon, Purity: ≥98%, CAS: 435-97-2, 50 mg
SDP
Catalog No. 502991606
Encompass_Preferred
An anticoagulant and vitamin K antagonist; inhibits the activity of wild-type VKOR (IC50 = 4.2 nM), as well as a variety of mutant VKORs (IC50s = 5.1-835 nM), in cell-based reporter assays; inhibits prothrombin complex synthesis in rats in a dose-dependent manner
Catalog No. 50-299-1606 Supplier Cayman Chemical Supplier No. 3173050 MG
An anticoagulant and vitamin K antagonist; inhibits the activity of wild-type VKOR (IC50 = 4.2 nM), as well as a variety of mutant VKORs (IC50s = 5.1-835 nM), in cell-based reporter assays; inhibits prothrombin complex synthesis in rats in a dose-dependent manner