Learn More
Medchemexpress LLC Phenserine | 101246-66-6 | 99.6% | 337.42 | 5 MG
SDP
Catalog No. 5000364458
Encompass_Preferred
Phenserine, a derivative of Physostigmine, is a potent, noncompetitive, long-acting, and selective AChE inhibitor. It reduces β-amyloid precursor protein (APP) and β-amyloid peptide (Aβ) formation, improving cognitive performance and attenuating the progression of Alzheimer's disease.
  • Potent, noncompetitive, long-acting, and selective AChE inhibitor
  • Reduces β-amyloid precursor protein (APP) and β-amyloid peptide (Aβ) formation
  • Improves cognitive performance
  • Attenuates the progression of Alzheimer's disease
  • For research use only
Catalog No. 50-003-64458 Supplier Medchemexpress LLC Supplier No. 5MGHY103374
Phenserine, a derivative of Physostigmine, is a potent, noncompetitive, long-acting, and selective AChE inhibitor. It reduces β-amyloid precursor protein (APP) and β-amyloid peptide (Aβ) formation, improving cognitive performance and attenuating the progression of Alzheimer's disease.
  • Potent, noncompetitive, long-acting, and selective AChE inhibitor
  • Reduces β-amyloid precursor protein (APP) and β-amyloid peptide (Aβ) formation
  • Improves cognitive performance
  • Attenuates the progression of Alzheimer's disease
  • For research use only