Learn More
Medchemexpress LLC HY-103374 10mg Medchemexpress, Phenserine CAS:101246-66-6 Purity:>98%
SDP
Catalog No. 501968092
Encompass_Preferred
Medchemexpress, HY-103374 10mg Phenserine CAS:101246-66-6 Phenserine ((-)-Eseroline phenylcarbamate) is a derivative of Physostigmine and is a potent, noncompetitive, long-acting and selective AChE inhibitor. Phenserine reduces β-amyloid precursor protein (APP) and β-amyloid peptide (Aβ) formation. Phenserine improves cognitive performance and attenuates the progression of Alzheimer's disease[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8092 Supplier Medchemexpress LLC Supplier No. HY10337410MG
Medchemexpress, HY-103374 10mg Phenserine CAS:101246-66-6 Phenserine ((-)-Eseroline phenylcarbamate) is a derivative of Physostigmine and is a potent, noncompetitive, long-acting and selective AChE inhibitor. Phenserine reduces β-amyloid precursor protein (APP) and β-amyloid peptide (Aβ) formation. Phenserine improves cognitive performance and attenuates the progression of Alzheimer's disease[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.