Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenserine (-)-eseroline phenylcarbamate | 101246-66-6 | MFCD00672748 | 99.6% | 337.42 g/mol | C20H23N3O2 | 1 ML
SDP

Catalog No. 5000244949
Encompass_Preferred

Phenserine is a research-grade acetylcholinesterase (AChE) inhibitor supplied as a ready-to-use 10 mM stock solution in DMSO (1 mL). It is commonly used in vitro to study cholinergic signaling and amyloid-β related pathways; reported purity is ~99.6% and molecular weight is 337.42 g/mol.

  • Ready-to-use 10 mM stock solution in DMSO.
  • Supplied as a 1 mL vial for immediate use in assays.
  • High reported purity suitable for research applications.
  • Useful for acetylcholinesterase inhibition and amyloid-β pathway studies.
  • Molecular formula C20H23N3O2 and molecular weight 337.42 g/mol.

Catalog No. 50-002-44949 Supplier Medchemexpress LLC Supplier No. HY10337410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenserine is a research-grade acetylcholinesterase (AChE) inhibitor supplied as a ready-to-use 10 mM stock solution in DMSO (1 mL). It is commonly used in vitro to study cholinergic signaling and amyloid-β related pathways; reported purity is ~99.6% and molecular weight is 337.42 g/mol.

  • Ready-to-use 10 mM stock solution in DMSO.
  • Supplied as a 1 mL vial for immediate use in assays.
  • High reported purity suitable for research applications.
  • Useful for acetylcholinesterase inhibition and amyloid-β pathway studies.
  • Molecular formula C20H23N3O2 and molecular weight 337.42 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.