Learn More
Cayman Chemical PhensuxImIde 10mg
SDP
Catalog No. 502937590
Encompass_Preferred
An anticonvulsant; inhibits MES- and pentylenetetrazole-induced seizures in mice (ED50s = 112 and 50 mg/kg, respectively); induces proteinuria and hematuria in rats at 1.25 mmol/kg
Catalog No. 50-293-7590 Supplier Cayman Chemical Supplier No. 2760110 MG
An anticonvulsant; inhibits MES- and pentylenetetrazole-induced seizures in mice (ED50s = 112 and 50 mg/kg, respectively); induces proteinuria and hematuria in rats at 1.25 mmol/kg