Learn More
Cayman Chemical Phenethyl 4-ANPP, Purity: ≥98%, 5 mg
SDP
Catalog No. 502940071
Encompass_Preferred
An analytical reference standard that is structurally similar to known opioids; intended for research and forensic applications
Catalog No. 50-294-0071 Supplier Cayman Chemical Supplier No. 283865 MG
An analytical reference standard that is structurally similar to known opioids; intended for research and forensic applications