Learn More
TARGETMOL CHEMICALS INC PHENTOLAMINE 50MG
SDP
Catalog No. 502576246
Encompass_Preferred
Also available in 5 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Phentolamine mesylate is an alpha-1 and alpha-2 selective adrenergic receptor antagonistpurity: 99%
Catalog No. 50-257-6246 Supplier TARGETMOL CHEMICALS INC Supplier No. T834850MG
Also available in 5 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Phentolamine mesylate is an alpha-1 and alpha-2 selective adrenergic receptor antagonistpurity: 99%