Learn More
Medchemexpress LLC Phentolamine hydrochloride | 73-05-2 | 99.9% | 1 ML
SDP
Catalog No. 5000342666
Encompass_Preferred
Phentolamine hydrochloride is an orally active adrenergic α receptor-blocking agent. It is intended for research use only.
  • Orally active adrenergic α receptor-blocking agent
  • Supplied as 10 mM in 1 mL of DMSO
  • Purity of 99.87%
Catalog No. 50-003-42666 Supplier Medchemexpress LLC Supplier No. HY12717A10MM/1ML
Phentolamine hydrochloride is an orally active adrenergic α receptor-blocking agent. It is intended for research use only.
  • Orally active adrenergic α receptor-blocking agent
  • Supplied as 10 mM in 1 mL of DMSO
  • Purity of 99.87%