Learn More
Medchemexpress LLC Phentolamine hydrochloride | 73-05-2 | 99.9% | 100 MG
SDP
Catalog No. 5000342487
Encompass_Preferred
Phentolamine hydrochloride is an orally active, competitive α-adrenergic receptor-blocking agent. It functions as an antagonist with Ki values of 1.6 nM for α1-adrenergic receptors and 9 nM for α2-adrenergic receptors, making it suitable for research into vasodilation.
  • Competitive α-adrenergic receptor antagonist
  • Exhibits Ki of 1.6 nM for α1-adrenergic receptors
  • Exhibits Ki of 9 nM for α2-adrenergic receptors
  • Causes concentration-dependent relaxation (0.01-100 μM)
  • Inhibits phenylephrine-induced contraction in vitro
  • Inhibits epinephrine-induced hypertension and prolonged hypotension in cats
  • Inhibits norepinephrine-induced blood pressure in cats
  • For research use only
Catalog No. 50-003-42487 Supplier Medchemexpress LLC Supplier No. HY12717A100MG
Phentolamine hydrochloride is an orally active, competitive α-adrenergic receptor-blocking agent. It functions as an antagonist with Ki values of 1.6 nM for α1-adrenergic receptors and 9 nM for α2-adrenergic receptors, making it suitable for research into vasodilation.
  • Competitive α-adrenergic receptor antagonist
  • Exhibits Ki of 1.6 nM for α1-adrenergic receptors
  • Exhibits Ki of 9 nM for α2-adrenergic receptors
  • Causes concentration-dependent relaxation (0.01-100 μM)
  • Inhibits phenylephrine-induced contraction in vitro
  • Inhibits epinephrine-induced hypertension and prolonged hypotension in cats
  • Inhibits norepinephrine-induced blood pressure in cats
  • For research use only