Learn More
Abcam Phentolamine hydrochloride, Non-selective alpha antagonist, 100MG
Catalog No. 502707263
Encompass_Preferred
MW 317.8 Da, Purity >98%. Potent, reversible, non-selective α antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7263 Supplier Abcam Supplier No. AB120791100MG
MW 317.8 Da, Purity >98%. Potent, reversible, non-selective α antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement