Learn More
Selleck Chemical LLC Phentolamine Mesylate S2038-100mg
SDP
Catalog No. 503109979
Encompass_Preferred
Phentolamine Mesylate(Phentolamine methanesulfonate) is a reversible and nonselective alpha-adrenergic receptor antagonist used for the prevention or control of hypertensive episodes
Catalog No. 50-310-9979 Supplier Selleck Chemical LLC Supplier No. S2038100MG
Phentolamine Mesylate(Phentolamine methanesulfonate) is a reversible and nonselective alpha-adrenergic receptor antagonist used for the prevention or control of hypertensive episodes