Learn More
TARGETMOL CHEMICALS INC Phentolamine mesylate 500MG
SDP
Catalog No. 502875929
Encompass_Preferred
Also available in 1 mL, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Phentolamine mesylate (Phentolamine methanesulfonate) is a nonselective alpha-adrenergic antagonist used to treat hypertension and hypertensive emergencies, pheochromocytoma, vasospasm of [RAYNAUD DISEASE] and frostbite, clonidine withdrawal syndrome, impotence, and peripheral vascular disease. Purity 99.9%
Catalog No. 50-287-5929 Supplier TARGETMOL CHEMICALS INC Supplier No. T1275500MG
Also available in 1 mL, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Phentolamine mesylate (Phentolamine methanesulfonate) is a nonselective alpha-adrenergic antagonist used to treat hypertension and hypertensive emergencies, pheochromocytoma, vasospasm of [RAYNAUD DISEASE] and frostbite, clonidine withdrawal syndrome, impotence, and peripheral vascular disease. Purity 99.9%