Learn More
Apexbio Technology LLC Phentolamine Mesylate 65-28-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13635.1
Encompass_Preferred
Phentolamine Mesylate (CAS 65-28-1) is a non-selective alpha-adrenergic antagonist that blocks both alpha 1- and alpha 2-adrenoreceptors By preferentially inhibiting presynaptic alpha 2-adrenoreceptors phentolamine increases the release of neurotransmitters such as noradrenaline leading to enhanced functional biological modulation (FBM) while also exerting effects on postsynaptic alpha 1-adrenoreceptors This dual antagonism makes phentolamine a valuable tool for investigating adrenergic signaling pathways neurotransmitter regulation and vascular tone in various biomedical research settings
Catalog No. 50-000-91515